Would you like to log in again?
Adapter, Straight Connecting with PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
3842-11
89082-802EA 273.2 USD InStock
89082-802
Adapter, Straight Connecting with PTFE Stopcock, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This straight connecting adapter has a PTFE stopcock plug and drip tip in lower position.
  • PTFE stopcock plug
  • [ST] inner and reinforced outer joints
  • With drip tip in lower position
Adapter has a 4mm-bore 1:5 PTFE stopcock plug. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty