Would you like to log in again?
Adapter, Vacuum, Long Stem, Ace Glass Incorporated
Supplier: Ace Glass
9121-08 9121-04 9121-06
89082-826EA 130.82 USD InStock
89082-826 89064-996 89082-822
Adapter, Vacuum, Long Stem, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This apparatus is useful as a vacuum adapter or addition tube.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • With hose connection
  • Fabricated from borosilicate glass
The stem length below the joint on this adapter is 130mm. Use with a 5/16-inch I.D. tubing, size A hose connection. Adapter has an outer standard taper joint at the top and an inner standard taper joint at the bottom.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty