Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Glass or PTFE Baffles, Ace Glass Incorporated
Supplier: Ace Glass
12177-152 12177-154 12177-145 12177-148 12177-13 12177-10 12177-23 12177-19 12177-157
89427-772EA 1509.96 USD InStock
89427-772 89082-848 89082-850 89427-770 89427-768 89427-764 89082-846 89427-766 89082-844
Glass or PTFE Baffles, Ace Glass Incorporated
Stirrers Stirring Rods
Use Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint
Baffles can be adjusted vertically or rotated to increase agitation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty