Would you like to log in again?
Beakers, Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
5340-125 5340-103 5340-110 5340-120 5340-118 5340-130 5340-105 5340-135 5340-115
89082-872EA 237.47 USD InStock
89082-872 89082-882 89082-870 89082-880 89082-876 89082-886 89082-874 89082-884 89082-878
Beakers, Jacketed, Ace Glass Incorporated
Beakers
This jacketed beaker has one upper and one lower Ace-Thred connection on opposite sides.
Does not have a pouring spout. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty