Would you like to log in again?
Bearing/Debris Trap, PTFE, 19 mm, Ace Glass Incorporated
Supplier: Ace Glass
8067-59 8067-54 8067-09 8067-57 8067-07
89082-938EA 862.06 USD InStock
89082-938 89082-942 89084-584 89082-940 89084-926
Bearing/Debris Trap, PTFE, 19 mm, Ace Glass Incorporated
Stirrers Stirring Tools
Combination all-in-one stirring shaft bearing and debris trap
  • Complete item supplied with glass body, inner PTFE bearing, PTFE bushing and lock nut, Rulon saddle o-ring, “Ace-Safe” connector for 3/8-inch tubing, and PTFE plug.
Complete item supplied with glass body, inner PTFE bearing, PTFE bushing and lock nut, Rulon saddle o-ring, “Ace-Safe” connector for 3/8-inch tubing, and PTFE plug.
Designed to prevent particles from entering the reaction vessel. Lubricant-free PTFE bearing with compression saddle o-ring makes a mechanical seal against the polished glass shaft. Debris trap section consists of a PTFE washer and particle well. The #11 Ace-Thred on the borosilicate glass body side allows access for clean-out, or may be used for evacuation/gas purge port.
Ordering information: For replacement items, order as follows: inner bearing with o-ring (89084-584); glass filled PTFE bushing (89061-894); glass filled PTFE lock nut (89061-898); compression saddle with o-ring (89061-900); PTFE washer (89084-926); PTFE #7 front-seal plug (89052-846); #7 "Ace-Safe" connector (89052-946); #25 Ace-Thred glass adapters (45/50 = 89084-806; 55/50 = 89084-808; 71/60 = 89084-810).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty