Would you like to log in again?
316 Stainless Steel Stirrer Blades for 10 mm Button Shafts, Ace Glass Incorporated
Supplier: Ace Glass
8096-74 8096-72 8096-04 8096-10 8096-06 8096-70
89083-000EA 69.23 USD InStock
89083-000 89083-002 89083-004 89082-996 89082-994 89082-998
316 Stainless Steel Stirrer Blades for 10 mm Button Shafts, Ace Glass Incorporated
Stirrers Stirring Tools
The 316 stainless steel stir blades for 10 mm O.D
button type stir shafts are football shaped. Holed versions provide even greater agitation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty