Would you like to log in again?
Bottle, Threaded Neck, Ace Glass
Supplier: Ace Glass
4048-09 4048-45 4048-19 4048-13
89083-010EA 1011.21 USD InStock
89083-010 89083-012 89083-014 89083-008
Bottle, Threaded Neck, Ace Glass
Bottles Glass Bottles
This is a borosilicate glass bottle designed for the storage of solutions.
Bottles have a GL120 external thread finish neck with large 106 mm (4.2") Int.Ø opening. The 9.5 and 13.25 liter sizes have a conventional bottle shape. The 19 and 45.5 liter sizes are similar in design to a carboy.
Ordering information: A glass-filled PTFE cap with CAPFE (PTFE encapsulated silicone) O-ring may be purchased separately. Ask for VWR Order Code 89059-794.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty