Would you like to log in again?
'V' Assembly for Rotary Evaporators, Ace Glass Incorporated
Supplier: Ace Glass
3951-10 3951-05
89083-088EA 1533.07 USD InStock
89083-088 89083-092
'V' Assembly for Rotary Evaporators, Ace Glass Incorporated
Condensers
Complete "V" Assembly glassware set for many popular bench-scale rotary evaporators
Fabricated from borosilicate glass. Fits models 200/205 and Series 114-144. Available poly coated for safety, or uncoated. Includes the main glass condenser body with inner coil for vertical installation and heavy wall 1L receiving flask. The bottom condenser joint is spherical [SJ] 35/20 ball joint. The included round-bottom receiving flask has a [SJ] 35/20 socket joint. Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks.
Ordering information: Includes o-ring, 89059-774 GL thread caps, and 89059-810 angled hose connectors. The bottom joint is spherical [SJ] 35/20.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty