Would you like to log in again?
Tubing Connector with Quick Connect/Disconnect, Ace Glass Incorporated
Supplier: Ace Glass
12517-40 12517-14 12517-08
89083-350EA 29.81 USD InStock
89083-350 89083-352 89083-354
Tubing Connector with Quick Connect/Disconnect, Ace Glass Incorporated
Fittings and Connectors
This is a two-piece tubing connector, used to connect tubing from a pressure source to the manifold of the ACE pressure reactor
Offers the convenience of a quick connect/disconnect coupling. Body has MPT thread for connecting to an adapter at end of ACE pressure manifold. Coupling insert is connected to tubing from pressure source. Push coupling insert into coupling body for leak-tight connection; press thumb latch to quick disconnect. Fabricated of polypropylene.
Ordering information: Order body and insert separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty