Would you like to log in again?
Cloning Cylinder, Ace Glass Incorporated
Supplier: Ace Glass
3865-06 3865-52 3865-55 3865-08 3865-10
89083-360CS 137.7 USD InStock
89083-360 89083-362 89083-364 89083-356 89083-358
Cloning Cylinder, Ace Glass Incorporated
Cylinders Cloning Cylinders
You can clone a single cell or group of cells by surrounding them with this glass cylinder
Note: 89083-364 and 89083-358 are for use in the Disinfectant Procedure described in the 1992 J.A.O.A.C. Hard Surface Carrier Test for Efficacy Testing of Disinfectants: Collaborative Study. These tubes are glazed on both ends.
Dip the end of the cylinder into a sterile silicone grease before pressing to the bottom of a culture flask to create an isolated well.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty