Would you like to log in again?
DynaBloc Heating Blocks, Cylindrical, Ace Glass
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
13698-14 13698-49 13698-21 13697-04 13698-41 13696-07 13697-16 13698-12 13697-08 13698-05 13696-11 13699-09 13699-06 13697-40 13698-03 13699-04 13699-03 13698-37 13698-20 13698-13 13696-03 13698-30 13698-38
89083-408EA 619.33 USD InStock
89083-408 89083-400 89083-378 89083-366 89083-388 89083-402 89083-368 89083-404 89083-406 89083-370 89097-314 89083-392 89083-380 89083-394 89083-372 89083-382 89083-396 89083-374 89083-384 89083-376 89083-398 89083-386 89083-390
DynaBloc Heating Blocks, Cylindrical, Ace Glass
Block Heaters Blocks
These anodized aluminum blocks are ideal for heating and mixing solutions in a variety of vials, tubes, and flasks for applications such as digestion, extraction, distillation, and synthesis
Universal base plate is required for operating all DynaBloc cylindrical heating blocks and fits a standard, circular, 135mm diameter stirrer/hotplate top. Blocks can be separated from the base for easy cleaning and handling. The base has a hole in the side to accommodate a standard thermocouple sensor for heat measurement. Base plate dimensions: 135I.D.x164O.D.mm (55/16x67/16"). Blocks for vials and tubes have pre-drilled openings for standard-size glass vials and tubes, including small dram vials, chromatography vials, scintillation vials, EPA vials, VOC vials, and standard test tubes.
Blocks for flasks are machined to accommodate small round-bottom flasks and small Erlenmeyer flasks and include one hole to accommodate the extension rods and T-handle extractor. Clamp devices are necessary to stabilize glassware while in blocks.
Segmented blocks feature three sections with different sized holes per section. Choose different sized vials, tubes, or a flask to run simultaneously.
Accessories include clamp devices, a short support rod is for supporting flasks in the flask blocks, and a long support rod for condensers and other taller glass apparatus. T-handle extractor allows easy removal of the block even when hot.
System is easy to set up, allows switching from vials to flasks in seconds, and is easy to disassemble for cleaning. The configuration of the vial/flask holes provides optimum heating and mixing in every well. All blocks have a pre-drilled hole to accommodate a sensor probe and another hole for the T-handle extractor.
Ordering information: Base plates are required for all blocks. Clamp devices are required to stabilize flasks and condensers while in blocks. Support rods may be also be necessary for flasks, condensers, and other taller glass apparatus. Base plate, block, and accessories must be ordered separately. Instatherm blocks are for use in 89075-328 Instatherm bath (1.2 L).
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty