Would you like to log in again?
Round-Bottom Three-Neck Flasks, Angled Side Necks, Ace Glass Incorporated
Supplier: Ace Glass
6948-76 9465-09 6948-16 6948-104 6948-07 6948-81 6948-116 6948-05 9466-12 6948-112 6948-26 6948-70 6948-22 6948-67 9465-13 6948-108 6948-89 6948-20 6948-85 9466-08 6948-40 6948-80 6948-88 6948-32 6948-91 6948-110 9465-25 9465-29 6948-106 6948-72 6948-82 6948-10 9465-27 9466-06 6948-25 6948-58 6948-34 9466-04 6948-86 6948-02 9465-07 6948-75 6948-03 9465-36 6948-79 9465-05 6948-87 9465-11 6948-114 9466-10 6948-12 6948-78 6948-90 9465-34 6948-38 6948-92 6948-83 6948-24 6948-69 6948-71 9465-23 9466-14
89074-740EA 157.2 USD InStock
89074-740 89083-902 89083-904 89057-146 89074-744 89057-144 89074-742 89056-818 89074-748 89074-702 89074-746 89074-706 89056-466 89056-900 89056-902 89057-182 89056-866 89074-752 89056-908 89074-750 89074-756 89056-906 89083-894 89083-896 89074-758 89074-718 89056-870 89056-912 89056-914 89056-910 89056-832 89074-722 89056-916 89056-918 89074-688 89074-726 89083-888 89056-884 89056-922 89056-804 89056-924 89056-920 89083-914 89057-174 89083-916 89056-806 89056-926 89057-254 89056-808 89057-176 89056-928 89074-738 89074-736 89056-894 89056-890 89056-812 89056-814 89056-858 89056-930 89056-896 89056-932 89056-898
Round-Bottom Three-Neck Flasks, Angled Side Necks, Ace Glass Incorporated
Flasks Round Bottom Flasks
These round-bottom flasks have angled side necks and [ST] outer joints
The outer joints are tooled with reinforcement rings (full-sized joints only) for added strength and stability. The flare-out directly below the center joint facilitates cleaning of the flask and enables stirring blades to be easily removed. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty