Would you like to log in again?
Egg-Shaped Magnetic Spinbars, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13663-13 13663-09 13663-07 13663-11 13663-03 13663-05
89084-206EA 32.6 USD InStock
89084-206 89084-216 89084-202 89084-212 89084-222 89084-220
Egg-Shaped Magnetic Spinbars, PTFE, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
These egg-shaped stirring magnets provide their own pivot point without a separate ring or abrupt change in contour
They are designed specifically for use in round-bottom vessels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty