Would you like to log in again?
Spinwedge Magnetic Stir Bar, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13665-02 13665-04 13665-08 13665-06
89084-228EA 47.02 USD InStock
89084-228 89084-226 89084-224 89084-230
Spinwedge Magnetic Stir Bar, PTFE, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
Wedge-shaped PTFE magnetic stir bars are designed to “plow” up sediment for thorough dispersion
Provides very strong magnetic action. Ideal for viscous solutions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty