Would you like to log in again?
Single Tube Vacuum Manifolds, O-Ring Joint Connections and Glass Stopcocks, Ace Glass Incorporated
Supplier: Ace Glass
8740-20 8740-18 8740-05 8740-03 8740-22 8740-07
89084-248EA 784.28 USD InStock
89084-248 89084-266 89084-254 89084-242 89084-272 89084-260
Single Tube Vacuum Manifolds, O-Ring Joint Connections and Glass Stopcocks, Ace Glass Incorporated
Manifolds
The no.15 o-ring joint connections at each end
Three or four ports with 4 mm bore glass high vacuum stopcocks and standard taper joints or #15 o-ring joint connections off each port. The distance between ports is 95 mm, which will accept 250 mL flasks or smaller. Requires all stainless steel, pinch type screw locking clamp 89060-256 (for 15 o-ring joint).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty