Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Single Tube Vacuum Manifold, 35/20 Ball Joint Connections and Glass Stopcocks, Ace Glass Incorporated
Supplier: Ace Glass
8743-08 8743-06 8743-17 8743-04 8743-15 8743-19
89084-268EA 1008.41 USD InStock
89084-268 89084-256 89084-244 89084-274 89084-262 89084-250
Single Tube Vacuum Manifold, 35/20 Ball Joint Connections and Glass Stopcocks, Ace Glass Incorporated
Manifolds
With a spherical 35/20 ball joint connections at each end
Three or four ports with 4 mm bore glass high-vacuum stopcocks and standard taper joints or no. 15 o-ring joint connections off each port. The distance between ports is 95 mm, which will accept 250 ml flasks or smaller. Requires all stainless steel, pinch type screw locking clamp 89060-256 (for no.15 o-ring joint).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty