Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Double Tube Vacuum Manifold, O-Ring Joint Connections and Glass Stopcocks, Ace Glass Incorporated
Supplier: Ace Glass
8737-06 8737-24 8737-22 8737-02 8737-04 8737-20
89084-324EA 2212.31 USD InStock
89084-324 89084-312 89084-322 89084-310 89084-320 89084-308
Double Tube Vacuum Manifold, O-Ring Joint Connections and Glass Stopcocks, Ace Glass Incorporated
Manifolds
The no.15 o-ring joint connections at each end
Three or four ports with 4 mm bore glass double oblique stopcocks and standard taper joints or no.15 o-ring joint connections off each port. The distance between ports is 210 mm, which will accept 3 L flasks or smaller. Requires all-stainless-steel pinchtype screw locking clamp 89060-256.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty