Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Double Tube Vacuum Manifold with Glass Stopcocks, Ace Glass Incorporated
Supplier: Ace Glass
8738-14 8738-18 8738-44 8738-42 8738-16 8738-40
89084-328EA 2184.92 USD InStock
89084-328 89084-316 89084-326 89084-314 89084-330 89084-318
Double Tube Vacuum Manifold with Glass Stopcocks, Ace Glass Incorporated
Manifolds
With a spherical joint 35/20 ball joint at one end, a standard taper 24/40 outer standard taper joints at opposite end
Three or four ports with 4 mm bore glass double oblique stopcocks and standard taper joints or a no.15 o-ring joint connections off each port. The distance between ports is 210 mm, which will accept 3 l flasks or smaller. Requires all stainless steel, pinch type screw locking clamp 89060-256 (for no.15 o-ring joint).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty