Would you like to log in again?
Unjacketed Benchtop Reactor, All-In-One, Ace Glass Incorporated
Supplier: Ace Glass
6542-25 6542-12
89084-398EA 9787.51 USD InStock
89084-398 89055-512
Unjacketed Benchtop Reactor, All-In-One, Ace Glass Incorporated
Laboratory Reactor Systems
A complete, general purpose benchtop reactor.
  • All wetted surfaces are glass or PTFE
  • A sturdy stand and all necessary clamps and clips are included
Self-supported with a 20" x 15" footprint, 33" high. Reactor features a 100 mm(4") head with (4) 24/40 joints, an allihn condenser, a pressure equalizing funnel, overhead glass stirring assembly with variable speed motor and controller, heating mantle with type J PTFE covered probe and controller.
If working with pressure, We recommend the 89054-494 safety manifold.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty