Would you like to log in again?
Complete Photochemical Reaction System, Ace Glass Incorporated
Supplier: Ace Glass
7840-180 7840-360 7840-185 7840-175 7840-340 7840-320
89060-558EA 11265.21 USD InStock
89060-558 89084-402 89084-410 89060-556 89060-560 89084-406
Complete Photochemical Reaction System, Ace Glass Incorporated
Laboratory Reactor Systems
Complete reaction assembly with all parts needed for immediate operation
Borosilicate glass reactor has a standard taper 60/40 center joint, one- stadnard taper 14/20 angled joint for sparger tube, one-standard taper 24/40 vertical joint for condenser, and one-#7 Ace-Thred joint to accommodate thermometer. Volumes indicated are total volumes. Volume in reactive area of lamp is 40-50% of the total volume. Complete assembly consists of reactor, quartz immersion well, sparger tube, PTFE stir bar #7 thermometer bushing, three meters of 4.8 mm (3/16") I.D. PTFE tubing, stand, 450 watt lamp and 450 watt power supply.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty