Would you like to log in again?
Precision Seal™ Septa, Rubber, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Precision Seal™
9106-26 9106-139 9106-36 9106-35 9106-23 9106-124 9106-27 9106-128 9106-33 9106-22 9106-138 9106-37 9106-136 9106-129 9106-126 9106-140 9106-137 9106-122 9106-31 9106-29 9106-133 9106-24 9106-134 9106-21 9106-127 9106-120 9106-125 9106-131 9106-121 9106-19 9106-34 9106-119 9106-32 9106-135 9106-132 9106-38 9106-20 9106-123 9106-28 9106-130 9106-30 9106-25
89084-496PK 172.18 USD InStock
89084-496 89084-550 89084-494 89084-492 89084-490 89084-536 89084-558 89084-514 89084-512 89084-534 89084-556 89084-554 89084-510 89084-532 89084-498 89084-552 89084-530 89084-518 89084-538 89084-516 89084-540 89084-562 89084-560 89084-548 89084-526 89084-504 89084-568 89084-524 89084-546 89084-502 89084-544 89084-522 89084-500 89084-566 89084-488 89084-564 89084-520 89084-542 89084-486 89084-508 89084-506 89084-528
Precision Seal™ Septa, Rubber, Ace Glass Incorporated
Stoppers Rubber Stoppers
Seals are engineered for a precision fit (80% glass to rubber contact) in standard taper glassware joints and tubes
Precision-Seal™ septa are manufactured under “white room” conditions, from one certified raw material formulation, for absolute consistency in all sizes, lot-to-lot.
Ordering information: Available in red or white. Mixed set includes samples of each size available.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty