Seals are engineered for a precision fit (80% glass to rubber contact) in standard taper glassware joints and tubes
Precision-Seal™ septa are manufactured under “white room” conditions, from one certified raw material formulation, for absolute consistency in all sizes, lot-to-lot.
Ordering information: Available in red or white. Mixed set includes samples of each size available.