Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sparger with Easy Action Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
6452-07 6452-108 6452-03 6452-18 6452-16 6452-33 6452-132 6452-125 6452-14 6452-104 6452-106 6452-05 6452-123 6452-44 6452-145 6452-121
89084-614EA 508.72 USD InStock
89084-614 89084-636 89084-646 89084-612 89084-644 89084-622 89084-632 89084-654 89084-620 89084-598 89084-652 89084-596 89084-628 89084-606 89084-638 89084-604
Sparger with Easy Action Stopcock, Ace Glass Incorporated
Stirrers Stirring Tools
This is a sparger tube with an Ace fritted bottom available in various porosities
Top has either 0 – 3 or 0 – 5 Easy Action Stopcock.
Delivery information: The 90 degree side arm is available with a glass hose connection or a #7 Ace-Thred which promotes the use of our Ace-Safe connector (89052-936). The 10 mm O.D. spargers will fit inside #11 Ace-Threds, the 14 mm O.D. within #15 Ace- Threds. Threaded versions supplied with nylon bushing and fetfe o-ring (89051-122).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty