Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sparger, Straight with Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
6453-133 6453-21 6453-124 6453-06 6453-146 6453-23 6453-08 6453-126 6453-122 6453-109 6453-105 6453-25 6453-04 6453-32 6453-107 6453-45
89084-650EA 181.58 USD InStock
89084-650 89084-658 89084-624 89084-602 89084-656 89084-634 89084-600 89084-610 89084-642 89084-630 89084-640 89084-608 89084-618 89084-616 89084-626 89084-648
Sparger, Straight with Hose Connection, Ace Glass Incorporated
Stirrers Stirring Tools
Straight sparger tube with Ace fritted bottom available in various porosities
Top is either a 1/2" ID hose connection or a #7 Ace-Thred which allows the use of our Ace-Safe connector (89052-936). The 10 mm OD spargers will fit inside #11 Ace-Threds, the 14 mm OD within #15 Ace-Threds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty