Would you like to log in again?
Labjaws Buret Holder, Ace Glass Incorporated
Supplier: Ace Glass
11051-15
89084-682EA 504.17 USD InStock
89084-682
Labjaws Buret Holder, Ace Glass Incorporated
Clamps General Purpose Clamps
This buret support provides a large, stable work area, combining a 612 cm2 glazed porcelain base, for ample work space, with a double buret holder
The base is easy to clean, impervious to all ordinary reagents, and withstands high heat (up to 260°C). Weight is 2.3 Kg. The unit has four rubber feet to prevent sliding. It is supplied with a 60 cm high anodized aluminum support rod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty