Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Labjaws Lab Frame Set, Large, Ace Glass
Supplier: Ace Glass
11163-07
89084-706EA 1798.58 USD InStock
89084-706
Labjaws Lab Frame Set, Large, Ace Glass
Support Rods and Frames
Elaborate apparatus setups are possible with this set
Frame consists of (10) 5 cm rods, (3) 46 cm rods, (2) 91 cm rods, (10) 122 cm rods, (38) S connectors, (4) rod end connectors and (6) feet.
It can be assembled into a support as large as 122 cm square with a 46 cm base.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty