Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Labjaws Lab Frame Set, Medium, Ace Glass
Supplier: Ace Glass
11162-08
89084-710EA 1166.9 USD InStock
89084-710
Labjaws Lab Frame Set, Medium, Ace Glass
Support Rods and Frames
This set can be assembled into a support for medium sized labs in either a horizontal or vertical position
Frame consists of (8) 5 cm rods, (2) 46 cm rods, (5) 61 cm rods, (2) 91 cm rods, (4) 122 cm rods, (24) S connectors, (4) rod end connectors and (4) feet.
The base rods are 46 cm long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty