Would you like to log in again?
Labjaws Round Support Plate, Ace Glass
Supplier: Ace Glass
11172-17
89084-720EA 154.57 USD InStock
89084-720
Labjaws Round Support Plate, Ace Glass
Support Stands
This Labjaws aluminum plate supports apparatus with flat base on Flexaframe lattice
Support is 15 cm (6") in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty