Would you like to log in again?
Vacuum Traps, O-Ring Flange Connection, Ace Glass
Supplier: Ace Glass
8755-05 8755-13 8755-17 8755-10
89084-822EA 355.57 USD InStock
89084-822 89084-828 89084-826 89084-824
Vacuum Traps, O-Ring Flange Connection, Ace Glass
Vacuum Traps
This vacuum trap assembly has a Viton O-ring flange connection, offering flexibility and ease of assembly and disassembly for cleaning
This trap utilizes a stainless steel, locking pinch clamp. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty