Would you like to log in again?
Scientific Plastics LDPE Containment Tray, Ace Glass
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
13220-37 13220-28 13220-16 13220-33 13220-24 13220-26 13220-35 13220-39 13220-20 13220-22
89084-856EA 417.94 USD InStock
89084-856 89084-834 89084-854 89084-830 89084-852 89084-840 89084-838 89084-858 89084-836 76107-082
Scientific Plastics LDPE Containment Tray, Ace Glass
Spill Control Materials
These Containment Trays are made of rugged, molded Low Density Polyethylene (LDPE)
They are available with or without a drain spigot. Designed to place under a reactor or system to contain a spill or leak.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty