Would you like to log in again?
Photochemical Quartz Immersion Well, Low Temperature, Ace Glass Incorporated
Supplier: Ace Glass
7876-10 7876-50
89084-932EA 3605.84 USD InStock
89084-932 89084-930
Photochemical Quartz Immersion Well, Low Temperature, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Triple walled, quartz, immersion well for use in temperatures as low as –78 °C
Same as the immersion well, except, plain-sided, (without standard taper joint), for use with #50 Ace-Thred bushing and all vessels. 415 mm total jacket length. Will accommodate 89061-252 and 89061-256 inner tubes. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty