Would you like to log in again?
Double-Tube Vacuum or Inert Gas Manifold, Ace Glass Incorporated
Supplier: Ace Glass
8729-52
89085-060EA 1722.44 USD InStock
89085-060
Double-Tube Vacuum or Inert Gas Manifold, Ace Glass Incorporated
Manifolds
Greaseless, vacuum/gas manifold with 0 - 4 mm high vacuum and Easy-Action three-way PTFE stopcocks for takeoffs
The bottom and top manifold tubes each feature a #7 Ace-Thred connection for easy cleaning. The ends are supplied with nylon stopper/plugs with FETFE O-Rings (89052-828). The port connections are 8 mm (1/32") O.D. The vacuum and gas connections are 12.7 mm (1/2") O.D. The distance between takeoffs is 100 mm (315/16"). Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty