Would you like to log in again?
Volumetric Flasks with Wide Neck and [ST] Glass Stopper, Class A, Hirschmann
Supplier: Hirschmann Laborgeräte
282G50W 282G25W 282G100W
89085-144PK 126.5 USD InStock
89085-144 89085-140 89085-142
Volumetric Flasks with Wide Neck and [ST] Glass Stopper, Class A, Hirschmann
Flasks Volumetric Flasks
Flasks include a [ST] glass stopper, a fine white graduation line, and a marking spot
Meet ASTM E288, Class A requirements.
Flasks are designed with wide necks for easy pipet access. Graduated to contain. All flasks feature lot number and production year printed onto the vessel for QA purposes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty