Synonyms:
Ethyl alcohol
ET107-4LTGL
ET107-100ML
ET107-1LTGL-CS
ET107-500MLGL
ET107-500ML-CS
ET107-1LTGL
ET107-4LTGL-CS
75811-080CS
1310
USD
InStock
75811-080
89085-238
89085-244
89085-240
89085-242
75811-084
75811-086
Ethanol ≥99.5% (v/v) USP (200 Proof) (not denatured)
Ethanol
Dehydrated Alcohol, 200 Proof, Undenatured, USP is used as an antiseptic to disinfect skin prior to injections, an active ingredient for pain relief and as a solvent. Spectrum manufacturers Dehydrated Alcohol, 200 Proof, Undenatured, USP and packages it in sizes that various sizes from convenient laboratory 100 mL bottles to 200 L production drums. Like all Spectrum Chemical USP products, dehydrated alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Certifications: Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)