Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol ≥99.5% (v/v) USP (200 Proof) (not denatured)

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger
Synonyms: Ethyl alcohol

ET107-4LTGL ET107-100ML ET107-1LTGL-CS ET107-500MLGL ET107-500ML-CS ET107-1LTGL ET107-4LTGL-CS
75811-080CS 1310 USD InStock
75811-080 89085-238 89085-244 89085-240 89085-242 75811-084 75811-086
Ethanol ≥99.5% (v/v) USP (200 Proof) (not denatured)
Ethanol
Dehydrated Alcohol, 200 Proof, Undenatured, USP is used as an antiseptic to disinfect skin prior to injections, an active ingredient for pain relief and as a solvent. Spectrum manufacturers Dehydrated Alcohol, 200 Proof, Undenatured, USP and packages it in sizes that various sizes from convenient laboratory 100 mL bottles to 200 L production drums. Like all Spectrum Chemical USP products, dehydrated alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Certifications: Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)


Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 78 °C (1013 hPa)
Melting Pt: –114.5 °C
Density: 0.794
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
Order Now
Specification Test Results
Assay by volume 99.5% Min.
Organic impurities max 200 ppm
Methanol max 10 ppm
Benzene max 2 ppm
UV ABSORPTION
240 nm 0.40
250 - 260 nm 0.30
270 - 340 nm 0.10

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty