Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Beakers, Griffin Low Form, Ace Glass Incorporated
Supplier: Ace Glass
12420-06 12420-22 12420-20 12420-12 12420-18 12420-10 12420-16 12420-08 12420-14
89085-376CS 410.54 USD InStock
89085-376 89085-386 89085-378 89085-372 89085-382 89085-374 89085-384 89085-370 89085-380
Beakers, Griffin Low Form, Ace Glass Incorporated
Beakers
These Griffin-style polypropylene beakers are for general laboratory use
  • Autoclavable
  • Approximate raised volume scales
  • Tapered walls for safe handling and convenient stacking
Combines a “no-drip” pouring spout with unbreakability and maximum translucency.
Ordering information: Lids are not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty