Would you like to log in again?
NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NextGen™ V Vial®
W986339NG W986354NG W986288NG W986293NG W986336NG W986279NG W986359NG W986284NG W986261NG W986298NG W986287NG W986299NG W986273NG W986257NG W986289NG W986256NG W986283NG W986319NG W986253NG W986294NG W986254NG W986260NG W986281NG W986276NG W986282NG W986334NG W986277NG W986314NG W986356NG W986259NG W986290NG W986297NG W986274NG W986316NG W986272NG
89085-434CS 173.92 USD InStock
89085-434 89085-456 89085-458 89085-436 89085-452 89085-430 89085-496 89085-476 89085-454 89085-432 89085-416 89085-438 89085-418 89085-470 89085-492 89085-450 89085-472 89085-490 89085-424 89085-446 89085-448 89085-426 89085-486 89085-420 89085-442 89085-464 89085-422 89085-466 89085-444 89085-428 89085-460 89085-482 89085-440 89085-462 89085-480
NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton, DWK Life Sciences
Vials
Sturdy screw-thread vials have a conical interior bottom for maximum retrieval of content.
Vials can be used for small-scale reactions, centrifugation, storage, and shipping. They are ideal for use in the biochemical, pharmaceutical, and radiochemical industries. Fire-glazed bottom protects against glass particulates. Low-particulate packaging protects against contamination during transportation and storage. Clear vials are made from Wheaton 33* low extractable borosilicate glass to safeguard against a change in product pH caused by alkali, which can be extracted by ordinary glass. Amber vials are made from Wheaton 320 low extractable borosilicate glass. Caps are black, phenolic solid-top or open-top with fluoropolymer resin-faced 14B rubber liners.
Certifications: Clear vials conform to ASTM Type I, Class A, and USP Type I requirements. Amber vials conform to USP Type I requirements.
Ordering information: These vials do not have interior ground joints.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty