Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Deepwell Plates 96-well, 1000 µl

Supplier: Eppendorf
951033103 951032760 0030113535 951032689 951032662 951032701 951033006 951032727 951033103 951032786 951032701 951033006 951032603 951032603 0030113535 951032620
77395-522PK 173.68 USD InStock
CA89085-632 77395-522 89085-632 CA89085-630 89087-594 89087-600 89087-586 89087-598 89085-630 CA89085-628 89085-626 CA89085-626 89085-628 CA77395-522 89087-592 89087-590
Deepwell Plates 96-well, 1000 µl
Microplates Deep Well Microplates
Eppendorf Deepwell plates 96-well, 1000 µl offer efficient sample handling with features like OptiTrack® matrix for rapid identification and g-Safe® technology for stability during centrifugation. SafeCode variants provide secure three-level coding, including error correction for reliable identification, ensuring flexibility and accuracy in experimental workflows.

  • OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
  • g-Safe®: Exceptional centrifugation stability up to 6000×g for faster protocols and improved sample quality
  • Raised well rims and smooth surface for reliable closing, even with heat sealing
  • Easy and reliable stacking
  • Minimized remaining volume and high uniformity from well to well
  • High-quality polypropylene ensures high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without slip agents, plasticizers, and biocides - materials which could have a negative effect on bioassays
  • Autoclavable 121 °C, 20 min

For more dependability, stability and efficiency choose Eppendorf deepwell plates. Available in both 96- and 384-well formats with white, yellow, green or blue borders, these deep well plates offer high-performance in all applications both manual and automated. With OptiTrack® matrix, a high-contrast alphanumeric label, laser-engraved in the border, you'll be able to find your sample up to 30% faster and reduce pipetting errors. Rely on Eppendorf deepwell plates for everything from sample storage at 86 °C to DNA denaturation at 100 °C, and set new standards in your laboratory.

Safe code option provides three-level coding for safe sample and vessel identification. ECC 200 error correction function enables safe code reading with up to 30% damaged codes.

Volume: 1000 µl
No. of wells: 96
Dimensions: 127.8×85.5×44.1 mm
Material: Polypropylene (PP)
Bottom shape: Conical

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty