Would you like to log in again?
Axygen® Boil-Proof Microcentrifuge Tubes, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
MCT-150-C-CC MCT-150-SPCC
89087-346CS 1006.54 USD InStock
89087-346 89087-348
Axygen® Boil-Proof Microcentrifuge Tubes, Corning
Tubes Microcentrifuge Tubes
Axygen's spectum color closed cap boil-proof tubes will not open during boiling.
Composed of a homopolymer, tubes have ultrasmooth conical bottoms to allow 100% visibility and no sharp 'gates' to cause cuts and scratches. Frosted side surface area for writing. Extremely accurate and visible graduation markings. Thin membrane in cap middle allows easy syringe penetration. Diamond polished molds allow tubes to be manufactured without contaminating releasing fluids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty