Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Traceable® GIANT-DIGITS™ Radio-Controlled Atomic Clocks

Supplier: Avantor
VWR® Traceable® Giant-Digits™
90007-95
89087-394EA 187.81 USD InStock
89087-394
VWR® Traceable® GIANT-DIGITS™ Radio-Controlled Atomic Clocks
Timers Wall Clocks
Digital clocks display time and temperature, with large digits that are readable from 61 m (200').

  • Traceable
  • Linked to atomic clock in Boulder, Colorado
  • Automatically adjusts for daylight savings and leap year
  • Ambient and remote temperatures are displayed in °C and °F

Clocks receive the correct time by radio signal from the NIST Colorado atomic clock.

Temperature accuracy: ±1 °C (±1.8 °F)
Temperature resolution: 0.1°

Time accuracy: 0.0001 s
Time resolution: 1 s

Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Ordering information: Supplied with batteries. Clock is supplied with a remote temperature module.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty