Would you like to log in again?
VWR® Traceable® Giant-Digit™ Countdown Timer
Supplier: VWR
VWR® Traceable® Giant-Digits™
90012-91
89087-398EA 38.49 USD InStock
89087-398
VWR® Traceable® Giant-Digit™ Countdown Timer
Timers Digital Timers
Timer can be programmed in countdown mode or stopwatch mode up to 99 minutes, 59 seconds.
  • Traceable
  • Memory function for repetitive timing
  • Magnetic back for mounting on metal surfaces
Timer features Automatic Bounceback™ memory for repetitive timing. Three labeled buttons for intuitive operation. Giant-digit 3.4 cm (1⁵/₁₆") display can be read from 15 m (50').
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Delivery information: Timer is supplied with battery, easel bench stand, and magnetic back.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty