Would you like to log in again?
VWR® Traceable® Multi-Colored™ Timer
Supplier: VWR
VWR® Traceable® Multi-Colored™
90012-98
89087-400EA 52.14 USD InStock
89087-400
VWR® Traceable® Multi-Colored™ Timer
Timers Digital Timers
Four separate timing channels can be programmed in countdown mode or stopwatch mode up to 99 hours, 59 minutes, 59 seconds.
  • Traceable
  • Four-channel alarm timer
  • Displays countdown alarm, stopwatch, or time-of-day
Timer features Automatic Bounceback™ memory for repetitive timing. Timer can also display time of day in 12- or 24-hour mode. Supplied with battery, easel bench stand, magnetic back, and three colorful frames to personalize the timer.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty