Would you like to log in again?
SP Bel-Art HPLC Reservoir Secondary Containers, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
F16956-0001 F16956-0003 F16956-0002
89087-452EA 384.23 USD InStock
89087-452 89087-450 89087-448
SP Bel-Art HPLC Reservoir Secondary Containers, Bel-Art Products, a part of SP
Boxes Storage Boxes
Safeguard expensive HPLC solvent reservoirs against damage or breakage while in use or during storage with these durable polyethylene HPLC Reservoir Secondary Containers.
  • Durable polyethylene construction
  • Chemically resistant
  • Available in a variety of sizes
These containers are resistant to most chemicals and are available in 5 l, 10 l and 20 l sizes to accommodate popular HPLC solvent reservoirs.
In addition to protecting the HPLC reservoirs, the containers serve as secondary containment to prevent solvent spills in the work area. The large, unobstructed opening allows easy viewing of the HPLC reservoir, and the round shape of the containers minimizes wasted space on the benchtop.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty