Would you like to log in again?
Finntip® Flex Filter Tips, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
94056710 94056510 94056580 94056520 94056980 94056970 94056810 94056380
89047-108CS 297.81 USD InStock
89047-108 89047-106 89047-114 89047-112 89049-826 89047-110 89087-482 89087-480
Finntip® Flex Filter Tips, Thermo Scientific
Pipette Tips
The Finntip Flex Filter Pipette Tips combine the ergonomics of a standard Finntip Flex pipette tip and the protection provided by a filtered tip.
Finntip flex filter pipette tips effectively minimize the risk of passing contamination from the pipette to the sample. Including a range of products certified free from DNA, DNase, RNase and endotoxin, these sterile tips suit all sensitive procedures, including PCR-based reactions where contamination is a concern. The filters are composed of an inert non-sealing polyethylene matrix and are effective at eliminating carryover contamination.
With the soft Flex tips, the amount of force needed to attach and eject tips is significantly reduced for enhanced pipetting ergonomics.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty