Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Laundry Bag, Associated Bag
Supplier: Associated Bag
19-1-05 19-1-07
89087-552CS 594.5 USD InStock
89087-552 89087-548
Laundry Bag, Associated Bag
Bags Laundry Bags
Perfect for hospitality and healthcare industries, these laundry bags are water soluble.
  • Puncture and static - resistant
  • Attached tie tap for closure
  • Cost effective, puncture-resistant bags hold wet or damp linens
They won’t damage fabrics, equipment, or waste-water treatment systems. The bags are highly resistant to most organic solvents, greases, fats, and oils and are impervious to most gases. They won't generate a static charge.
Bags dissolve completely in water at 140 °F and fit 25" hamper.
Packaging: 100 per Carton.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty