Would you like to log in again?
SP Bel-Art Lab-Aire® II Glove Holder, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F18931-0000 F189310000
89087-556EA 52.58 USD InStock
89087-556 CA95042-252
SP Bel-Art Lab-Aire® II Glove Holder, Bel-Art Products, a part of SP
Glove Dispensers and Glove Box Holders
Realize faster drying time by placing wet laboratory gloves on the convenient Lab-Aire® II Glove Holder.
  • Epoxy coating
  • Autoclavable
  • Great chemical and corrosion resistance
  • Holder extends glove life
  • Electric drying racks circulate heated air
This holder attaches easily to all Lab-Aire® II Dryers and keeps gloves open for external as well as internal drying
Significantly reduce drying time by attaching the glove holder to Electric Lab-Aire® II Drying Racks. The electric drying racks circulate heated air in and around the gloves. Glove holder is steam autoclavable at 121°C (250°F).
The holder helps to extend the life of gloves by maintaining their shape, and preventing bending, cracking, and moisture buildup. Each holder can hold one pair of gloves and is epoxy-coated for chemical and corrosion resistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty