Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
AzerMark™ Etching Slides, Azer Scientific
Supplier: Azer Scientific
AzerMark™
EMS300BW
89087-558CS 495.8 USD InStock
89087-558
AzerMark™ Etching Slides, Azer Scientific
Slides Microscope Slides
These slides are designed for use with automated slide etch labeling machines.
  • White glass with ground edges
  • Black and white labeling area
  • Dimensions: 75 x 25 x 1 mm
  • Adhesion available
The slides have white glass with ground edges, and the frosted coating retains most laboratory marking methods.
They are available with or without adhesion and include a black and white labeling area.
Ordering information: Slides are packaged 72 per box, 20 boxes per case (10 gross).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty