Would you like to log in again?
Eppendorf® 96-Well Deepwell Plates
Supplier: Eppendorf
Eppendorf®
951033103 951033481 951033421 951033707 951031984 951031925 951031861 951033464 951032883 951031909 951033405 951032760 951032301 951033600 951031909 951033502 951033600 951032301 951031828 951032689 951032662 951032701 951033707 951033006 951033405 951032727 951033103 951031801 951032085 951033308 951032883 951032786 951032085 951032701 951032123 951033529 951033006 951032204 951033561 951032204 951032603 951033502 951032603 951031801 951032921 951033588 951031887 0030504305 951031968 951032620
89085-632CS 711.88 USD InStock
89085-632 89087-640 CA89085-644 CA89085-642 CA89085-640 89087-600 89085-630 89087-642 89087-608 CA89085-648 CA89085-646 CA89087-608 89085-644 89085-646 CA89208-776 89087-594 89087-632 89085-642 89087-630 89087-598 CA89087-640 CA89085-618 89085-648 89087-592 89087-590 89087-584 CA89085-622 89087-582 CA89085-620 89087-622 89087-586 89087-626 89085-618 CA89085-628 89087-624 CA89085-626 CA89085-624 CA89085-632 89085-622 89087-574 CA89085-630 89085-624 89087-610 89087-578 89085-620 89087-576 89087-618 89085-626 89085-628 89087-570
Eppendorf® 96-Well Deepwell Plates
Microplates Deep Well Microplates
Eppendorf® sets the new standard for deep well plates with new RecoverMax well designs.
  • Made of virgin polypropylene, each plate type is available as standard and sterile quality
  • Ease of stacking, multiple sealing options as well as high chemical and temperature resistance make these plates excellent, multipurpose tools
The well geometry enables high-speed pipetting, excellent mixing and high-speed centrifugation to accelerate your manual as well as your automated work processes. Sophisticated colors and coding make reading, orientation and manual handling easier and faster.
Deep well Plate 96/500, 96/1000 and 384/200 are also available as DNA LoBind and Protein LoBind quality. Reliable centrifugation stability Eppendorf deep well plates 96 and 384 are designed to be centrifugation-stable and durable in a temperature range from 4 to 40 °C. Standard, DNA and Protein LoBind Eppendorf deep well Plate 96 and 384 can be centrifuged up to 6000×g². Sterile Eppendorf plate deep well 96 and 384 are centrifugation stable up to 5000×g².
For sample recovery after processing or storage, centrifuge 5804 R and 5810 R and rotors A-4-81-MTP and A-2-DWP are recommended.
Certifications: The plates fulfill current guidelines: SBS1, EN ISO 9001/13485.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty