Would you like to log in again?
VWR® Film Seals for Fluorescence and Photoprotection, Black
Supplier: Avantor
VWR®
BKNL50
89087-692CS 80.38 USD InStock
89087-692
VWR® Film Seals for Fluorescence and Photoprotection, Black
Seals Microplate Seals
These films are 240 µm thick black vinyl with adhesive for use with 96-well, 384-well, and 1536-well black microplates. They fit within the edges of raised-rim plates and may be applied to either the plate top or bottom.
  • Edge cutouts assist in positioning around ribs on the plate bottom
  • Split backing aids in applying the film properly
  • No end tabs
  • Recommended temperature range −40 to +80 °C (-40 to 176 °F)
The light-blocking and absorbing properties of these films make them ideal for reducing stray light and crosstalk in fluorescence applications and for protecting light-sensitive samples in storage.
Each film L×D: 114×76 mm
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty