Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® White Films for Luminescence and Microscopy

Supplier: VWR
VWR®
WTNL50
89087-696CS 51.86 USD InStock
89087-696
VWR® White Films for Luminescence and Microscopy
Seals Microplate Seals
These films are 240 µm thick white vinyl with adhesive, for use with white microplates.

  • Backing split on the long dimension aids in application of film to plates
  • Film can be applied to either top or bottom
  • No end tabs
  • Recommended temperature range −40 to +80 °C

Light reflective properties of the film, when used to seal clear-bottom plates, maximise the sensitivity of bioluminescence and chemiluminescence assays in bottom-reading Luminometers. White films have also been used as a backing matrix for microscopy of punch-out samples from filter plates.

Each film L×D: 116×76 mm (49/16×3").

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty